Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SMTN SIRNA Assays ›
Gene Symbol:
SMTN
Gene Name:
smoothelin
Gene Aliases:
-
Chromosome Location:
Chr.22: 31081296 - 31104624 on Build GRCh38
UniGene ID:
Hs.149098
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s12958
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SMTN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001207017.1
NP_001193946.1
13
2089
NM_001207018.1
NP_001193947.1
11
2018
NM_006932.4
NP_008863.3
12
2014
NM_134269.2
NP_599031.1
12
2014
NM_134270.2
NP_599032.2
12
2014
XM_006724298.1
XP_006724361.1
13
2085
XM_006724302.1
XP_006724365.1
13
2144
XM_011530333.2
XP_011528635.1
14
2190
XM_011530336.2
XP_011528638.1
14
2191
XM_011530337.2
XP_011528639.1
14
2191
XM_011530338.2
XP_011528640.1
13
2097
XM_011530339.2
XP_011528641.1
14
2191
XM_011530340.2
XP_011528642.1
13
2098
XM_011530341.1
XP_011528643.1
6
1022
XM_017028913.1
XP_016884402.1
14
2191
XM_017028914.1
XP_016884403.1
13
2097
XM_017028915.1
XP_016884404.1
13
2097
XM_017028916.1
XP_016884405.1
14
2191
XM_017028917.1
XP_016884406.1
14
2191
XM_017028918.1
XP_016884407.1
14
2191
XM_017028919.1
XP_016884408.1
13
2097
GenBank mRNA
AB209643.1
-
9
1587
AF064238.3
-
12
1991
AJ010306.2
-
12
1991
AK092684.1
-
2
660
AK095916.1
-
3
433
AK095930.1
-
3
546
AK096953.1
-
12
2049
AK128627.1
-
12
3652
AK128719.1
-
12
1860
AK225648.1
-
3
455
AK293681.1
-
3
388
AK295418.1
-
11
2018
AK300064.1
-
3
535
AK301289.1
-
2
290
AK304084.1
-
13
2089
AK307627.1
-
3
463
AK309502.1
-
3
527
AY061971.1
-
12
1992
AY061972.1
-
12
1992
BC034237.2
-
12
1982
3912887
Y13492.2
-
12
1992
Z49989.2
-
3
448
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
smooth muscle contraction
muscle organ development
actin binding
structural constituent of muscle
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0