Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012895.3 | NP_037027.2 | 8, 9 | 767 | ||
XM_006251631.1 | XP_006251693.1 | 8, 9 | 881 | |||
GenBank mRNA | BC081712.1 | - | 8, 9 | 767 | 7193044 | |
FQ209450.1 | - | 3, 4 | 236 | |||
FQ218369.1 | - | 4, 5 | 430 | |||
FQ219573.1 | - | 2, 3 | 75 | |||
FQ225093.1 | - | 4, 5 | 438 | |||
U90340.1 | - | 8, 9 | 770 |