Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cadps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013219.1 | NP_037351.1 | 1038 | |||
GenBank mRNA | U16802.1 | - | 1038 |