Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC20A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005415.4 | NP_005406.3 | 7 | 1575 | ||
XM_017004768.1 | XP_016860257.1 | 7 | 1324 | |||
XM_017004769.1 | XP_016860258.1 | 8 | 3802 | |||
XM_017004770.1 | XP_016860259.1 | 8 | 1605 | |||
XM_017004771.1 | XP_016860260.1 | 9 | 1641 | |||
GenBank mRNA | AK123420.1 | - | 2 | 1113 | ||
AK312738.1 | - | 7 | 1494 | |||
BC019944.1 | - | 7 | 1482 | 3918690 | ||
BC075818.1 | - | 1 | 84 | |||
BX647555.1 | - | 1 | 719 | |||
L20859.1 | - | 7 | 1406 |