Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMBN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016519.5 | NP_057603.1 | 13 | 1048 | ||
GenBank mRNA | AF209780.1 | - | 13 | 1030 | ||
AF219994.2 | - | 13 | 1012 | |||
AF263464.1 | - | 13 | 1022 | |||
AJ537437.1 | - | 13 | 1022 | |||
AK314918.1 | - | 13 | 983 | |||
BC106931.1 | - | 13 | 983 | 40033703 | ||
BC106932.1 | - | 13 | 980 | 40033705 |