Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017190.4 | NP_058886.1 | 8 | 1219 | ||
XM_006228824.2 | XP_006228886.1 | 7 | 1249 | |||
XM_006228825.2 | XP_006228887.1 | 7 | 1766 | |||
XM_006228827.2 | XP_006228889.1 | 8 | 1301 | |||
XM_008759137.1 | XP_008757359.1 | 7 | 1773 | |||
XM_008759138.1 | XP_008757360.1 | 7 | 1242 | |||
XM_008759139.1 | XP_008757361.1 | 7 | 1772 | |||
GenBank mRNA | M11721.1 | - | 2 | 135 | ||
M14871.1 | - | 7 | 1189 | |||
M16800.1 | - | 8 | 1219 | |||
M22357.1 | - | 7 | 1167 | |||
M36702.1 | - | 2 | 135 | |||
X05301.1 | - | 7 | 1182 |