Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003801.1 | NP_001003801.1 | 5 | 586 | ||
NM_001003802.1 | NP_001003802.1 | 6 | 575 | |||
NM_003078.3 | NP_003069.2 | 6 | 614 | |||
XM_011516521.2 | XP_011514823.1 | 5 | 693 | |||
XM_017012556.1 | XP_016868045.1 | 5 | 413 | |||
GenBank mRNA | AK127595.1 | - | 6 | 1122 | ||
AK297876.1 | - | 5 | 608 | |||
AK309500.1 | - | 5 | 571 | |||
AK316173.1 | - | 5 | 558 | |||
AY450430.1 | - | 5 | 890 | |||
AY450431.1 | - | 6 | 575 | |||
BC002628.2 | - | 6 | 575 | 3608512 | ||
BC110350.1 | - | 5 | 505 | 5212363 | ||
BI598614.1 | - | 6 | 625 | |||
BX648385.1 | - | 5 | 368 | |||
U66619.1 | - | 6 | 623 |