Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322902.1 | NP_001309831.1 | 7 | 714 | ||
NM_001322903.1 | NP_001309832.1 | 8 | 810 | |||
NM_001322904.1 | NP_001309833.1 | 8 | 780 | |||
NM_001322905.1 | NP_001309834.1 | 8 | 743 | |||
NM_001322906.1 | NP_001309835.1 | 8 | 737 | |||
NM_001322907.1 | NP_001309836.1 | 9 | 938 | |||
NM_001322908.1 | NP_001309837.1 | 9 | 861 | |||
NM_001322909.1 | NP_001309838.1 | 8 | 728 | |||
NM_001322910.1 | NP_001309839.1 | 8 | 835 | |||
NM_003081.4 | NP_003072.2 | 7 | 683 | |||
NM_130811.3 | NP_570824.1 | 7 | 683 | |||
XM_005260808.4 | XP_005260865.1 | 7 | 761 | |||
XM_017028021.1 | XP_016883510.1 | 8 | 613 | |||
XM_017028022.1 | XP_016883511.1 | 8 | 705 | |||
GenBank mRNA | AK026732.1 | - | 2 | 854 | ||
AK090857.1 | - | 2 | 1651 | |||
AK094560.1 | - | 8 | 769 | |||
AK098767.1 | - | 7 | 553 | |||
AK223617.1 | - | 7 | 645 | |||
AK289647.1 | - | 9 | 844 | |||
AK314359.1 | - | 7 | 594 | |||
BC010647.1 | - | 7 | 572 | 3867544 | ||
BI602062.1 | - | 7 | 659 | |||
BI604120.1 | - | 7 | 662 | |||
BT019684.1 | - | 6 | 440 | |||
BX647443.1 | - | 7 | 555 | |||
D21267.1 | - | 7 | 645 | |||
L19760.1 | - | 7 | 528 | |||
L19761.1 | - | 7 | 528 |