Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003083.3 | NP_003074.1 | 5 | 963 | ||
NR_030717.1 | 5 | 860 | ||||
GenBank mRNA | AK222926.1 | - | 5 | 934 | ||
AK314878.1 | - | 5 | 948 | |||
BC011868.2 | - | 5 | 918 | 4545178 | ||
BX647416.1 | - | 5 | 827 | |||
U44755.1 | - | 5 | 947 | |||
U44898.1 | - | 5 | 929 |