Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psma7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008217.1 | NP_001008218.1 | 2 | 256 | ||
GenBank mRNA | BC127466.1 | - | 2 | 276 | 7381383 | |
CF110813.1 | - | 2 | 210 | |||
CF976316.1 | - | 2 | 257 | |||
D30804.1 | - | 2 | 230 | |||
FQ226344.1 | - | 2 | 263 | |||
FQ231713.1 | - | 2 | 263 |