Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271045.1 | NP_001257974.1 | 2, 3 | 354 | ||
NM_001271046.2 | NP_001257975.1 | 2, 3 | 221 | |||
NM_017288.2 | NP_058984.1 | 2, 3 | 435 | |||
GenBank mRNA | AF182949.1 | - | 2, 3 | 215 | ||
BC094523.1 | - | 2, 3 | 354 | 7316686 | ||
FQ215379.1 | - | 1 | 12 | |||
FQ216374.1 | - | 2, 3 | 221 | |||
M91808.1 | - | 2, 3 | 434 |