Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003090.3 | NP_003081.2 | 2 | 201 | ||
NR_135508.1 | 2 | 201 | ||||
GenBank mRNA | AK223070.1 | - | 2 | 158 | ||
AK223110.1 | - | 2 | 123 | |||
AK290078.1 | - | 2 | 157 | |||
AK312200.1 | - | 2 | 132 | |||
AL538391.3 | - | 2 | 200 | |||
BC022816.1 | - | 2 | 154 | 4904205 | ||
BC067846.1 | - | 2 | 170 | 30330402 | ||
BC071717.1 | - | 1 | 109 | 4514975 | ||
X13482.1 | - | 2 | 153 |