Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003091.3 | NP_003082.1 | 3, 4 | 435 | ||
NM_198216.1 | NP_937859.1 | 3, 4 | 435 | |||
GenBank mRNA | AK301034.1 | - | 4, 5 | 491 | ||
AK301206.1 | - | 3, 4 | 369 | |||
AK312620.1 | - | 3, 4 | 370 | |||
AY742712.1 | - | 2, 3 | 272 | |||
BC080516.1 | - | 3, 4 | 353 | 2822685 | ||
BM559678.1 | - | 435 | ||||
CR456969.1 | - | 2, 3 | 272 | |||
J04564.1 | - | 3, 4 | 398 | |||
M34081.1 | - | 2, 3 | 251 | |||
M34082.1 | - | 2, 3 | 251 | |||
X15893.1 | - | 2, 3 | 272 | |||
X17567.1 | - | 3, 4 | 326 | |||
X17568.1 | - | 3, 4 | 311 |