Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CALM2 SIRNA Assays ›
Gene Symbol:
Calm2
Gene Name:
calmodulin 2
Gene Aliases:
Cam
Chromosome Location:
Chr.6: 11064750 - 11080134 on Build Rnor_6.0
UniGene ID:
Rn.5968
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s132299
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Calm2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
BC058485.1
-
244
6921326
FQ211494.1
-
244
FQ211970.1
-
240
FQ212357.1
-
241
FQ212579.1
-
240
FQ212640.1
-
240
FQ212691.1
-
240
FQ212789.1
-
244
FQ212869.1
-
244
FQ213028.1
-
240
FQ213029.1
-
238
FQ213091.1
-
240
FQ213115.1
-
246
FQ213200.1
-
243
FQ213216.1
-
240
FQ213310.1
-
133
FQ213366.1
-
239
FQ213496.1
-
243
FQ213520.1
-
244
FQ213526.1
-
240
FQ213581.1
-
240
FQ213598.1
-
240
FQ213637.1
-
240
FQ213695.1
-
240
FQ213703.1
-
240
FQ213760.1
-
240
FQ213762.1
-
245
FQ213913.1
-
240
FQ213941.1
-
244
FQ213981.1
-
241
FQ213985.1
-
134
FQ213999.1
-
239
FQ214059.1
-
244
FQ214181.1
-
244
FQ214262.1
-
244
FQ214491.1
-
240
FQ214500.1
-
240
FQ214519.1
-
240
FQ214525.1
-
240
FQ214540.1
-
244
FQ215159.1
-
244
FQ215317.1
-
240
FQ215600.1
-
244
FQ216397.1
-
240
FQ216471.1
-
241
FQ218091.1
-
240
FQ220250.1
-
240
FQ220325.1
-
240
FQ220698.1
-
244
FQ220779.1
-
240
FQ222184.1
-
243
FQ223357.1
-
244
FQ225272.1
-
240
FQ226994.1
-
244
FQ229228.1
-
240
FQ229580.1
-
244
FQ229619.1
-
240
FQ229790.1
-
240
FQ231022.1
-
244
FQ231702.1
-
244
FQ233337.1
-
243
FQ234377.1
-
244
FQ235266.1
-
241
M17069.1
-
242
M19312.1
-
223
Back To Top
More Information
Panther Classification:
Molecular Function -
calmodulin-related
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0