Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004597.6 | NP_004588.1 | 3 | 368 | ||
NM_177542.3 | NP_808210.2 | 3 | 332 | |||
GenBank mRNA | AJ707552.1 | - | 1 | 53 | ||
AK291912.1 | - | 2 | 147 | |||
BC000486.2 | - | 2 | 139 | 2819767 | ||
BC001930.2 | - | 1 | 117 | 2819767 | ||
BG528418.1 | - | 263 | ||||
CB136390.1 | - | 2 | 558 | |||
CF130080.1 | - | 3 | 368 | |||
U15008.1 | - | 2 | 143 |