Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317165.1 | NP_001304094.1 | 4, 5 | 418 | ||
NM_001317166.1 | NP_001304095.1 | 3, 4 | 308 | |||
NM_001317167.1 | NP_001304096.1 | 3, 4 | 229 | |||
NM_001317168.1 | NP_001304097.1 | 3, 4 | 941 | |||
NM_001317169.1 | NP_001304098.1 | 4, 5 | 354 | |||
NM_001317171.1 | NP_001304100.1 | 2, 3 | 288 | |||
NM_003096.3 | NP_003087.1 | 3, 4 | 311 | |||
GenBank mRNA | AV682126.1 | - | 3, 4 | 311 | ||
BC000070.2 | - | 1 | 244 | 3503973 | ||
BC022432.1 | - | 3, 4 | 277 | 4276853 | ||
BC035103.1 | - | 3, 4 | 922 | 5262406 | ||
BC066302.1 | - | 1 | 234 | 30336948 | ||
BC070166.1 | - | 1 | 227 | 5809503 | ||
BC071880.1 | - | 1 | 201 | 3640165 | ||
BC106055.1 | - | 3, 4 | 294 | 4367206 | ||
BP424287.1 | - | 4, 5 | 358 | |||
BU540400.1 | - | 3, 4 | 277 | |||
CD109295.1 | - | 2, 3 | 257 | |||
CR456918.1 | - | 3, 4 | 189 | |||
HY168189.1 | - | 4, 5 | 355 | |||
X85373.1 | - | 3, 4 | 272 |