Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpsab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019322.2 | NP_062195.2 | 3 | 600 | ||
GenBank mRNA | BC061983.1 | - | 3 | 582 | 5598863 | |
U67910.1 | - | 3 | 599 |