Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019337.1 | NP_062210.1 | 4 | 307 | ||
XM_006230325.2 | XP_006230387.1 | 4 | 348 | |||
XM_006230326.2 | XP_006230388.1 | 4 | 360 | |||
GenBank mRNA | CD372843.1 | - | 4 | 307 | ||
U32437.1 | - | 2 | 79 |