Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209277.1 | - | 10 | 1732 | ||
AK302825.1 | - | 2 | 508 | |||
AK308978.1 | - | 4 | 756 | |||
AK309123.1 | - | 1 | 416 | |||
BC117261.1 | - | 10 | 1668 | 40125812 | ||
BC143366.1 | - | 10 | 1668 | 9051871 | ||
BC143367.1 | - | 9 | 1569 | 9051872 | ||
L13858.1 | - | 10 | 1642 |