Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022675.1 | NP_073166.1 | 3 | 120 | ||
XM_006239860.2 | XP_006239922.1 | 2 | 259 | |||
GenBank mRNA | D86642.1 | - | 3 | 120 |