Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request:org.apache.struts2.dispatcher.StrutsRequestWrapper@5e5906fa
Request attributes: pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
Popular
Applications & Techniques
Shop All Products
Services
Support
Sign in
Quick Order
Search
Thermo Fisher Scientific
Search All
Search
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
Attention: Existing favorites lists will be migrated to Shared Lists by the end of 2020.
[Learn more about Shared Lists]
LOADING
See other SOX5 SIRNA Assays ›
Gene Symbol:
SOX5
Gene Name:
SRY-box 5
Gene Aliases:
L-SOX5, L-SOX5B, L-SOX5F, LAMSHF
Chromosome Location:
Chr.12: 23529499 - 24562701 on Build GRCh38
UniGene ID:
Hs.657542
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s13303
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price (USD)
313.00
Your Price (USD)
Online offer (USD):
313.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SOX5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001261414.1
NP_001248343.1
17
2227
NM_001261415.1
NP_001248344.1
15
2267
NM_006940.4
NP_008871.3
15
2243
NM_152989.3
NP_694534.1
18
2476
NM_178010.2
NP_821078.1
7
1043
XM_011520831.2
XP_011519133.1
15
2105
XM_011520832.2
XP_011519134.1
15
2217
XM_011520833.2
XP_011519135.1
15
2546
XM_011520834.2
XP_011519136.1
19
2679
XM_011520835.2
XP_011519137.1
18
2731
XM_011520837.2
XP_011519139.1
15
2288
XM_011520838.2
XP_011519140.1
15
2108
XM_011520842.2
XP_011519144.1
8
1168
XM_017019888.1
XP_016875377.1
16
2560
XM_017019889.1
XP_016875378.1
16
2556
XM_017019890.1
XP_016875379.1
19
2695
XM_017019891.1
XP_016875380.1
19
2553
XM_017019892.1
XP_016875381.1
20
2774
XM_017019893.1
XP_016875382.1
17
2634
XM_017019894.1
XP_016875383.1
15
4564
XM_017019895.1
XP_016875384.1
15
2131
XM_017019896.1
XP_016875385.1
19
2692
XM_017019897.1
XP_016875386.1
19
2590
XM_017019898.1
XP_016875387.1
19
2587
XM_017019899.1
XP_016875388.1
18
2299
XM_017019900.1
XP_016875389.1
15
2913
XM_017019901.1
XP_016875390.1
15
7369
GenBank mRNA
AB081588.1
-
18
2474
AB081589.1
-
15
2243
AK074317.1
-
15
2170
AK098610.1
-
7
1043
AK223536.1
-
7
1043
AK303970.1
-
15
2267
BC029220.2
-
6
981
BC035227.2
-
6
981
BC047665.2
-
6
981
BC060773.1
-
17
2228
30343519
CR456713.1
-
6
893
JX570584.1
-
16
2288
JX570585.1
-
16
2183
S83308.1
-
6
977
Back To Top
More Information
Panther Classification:
Molecular Function -
HMG box transcription factor
nucleic acid binding
transcription factor
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-srch-1-prod-green:8080/10.253.225.215:8080.
git-commit: 0f6c802f6da5e59cd1b986a4dc444b4bcf3ac46d
git-url: http://victoria.invitrogen.com:8333/magellan/core.git
git-branch: origin/release/1.15.0-2021.01.02-1.0