Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022209.3 | NP_071545.2 | 3 | 445 | ||
GenBank mRNA | AY251277.1 | - | 3 | 397 | ||
BC078834.1 | - | 6 | 654 | 7114294 | ||
D14421.1 | - | 3 | 220 | |||
D38260.1 | - | 3 | 240 | |||
FM080850.1 | - | 3 | 445 |