Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npffr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022291.2 | NP_071627.2 | 1 | 241 | ||
XM_006256485.2 | XP_006256547.1 | 1 | 387 | |||
GenBank mRNA | AB040103.1 | - | 1 | 233 | ||
AF268901.1 | - | 1 | 233 | |||
AF330056.1 | - | 1 | 233 |
Set Membership: |
G-protein coupled receptor |