Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpbpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172073.1 | NP_742070.1 | 3 | 223 | ||
GenBank mRNA | AB009890.1 | - | 3 | 223 |