Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022596.2 | NP_072118.2 | 9, 10 | 707 | ||
XM_006233992.2 | XP_006234054.1 | 10, 11 | 757 | |||
XM_006233993.2 | XP_006234055.1 | 10, 11 | 757 | |||
XM_006233994.2 | XP_006234056.1 | 10, 11 | 757 | |||
XM_006233995.2 | XP_006234057.1 | 9, 10 | 676 | |||
XM_006233996.2 | XP_006234058.1 | 9, 10 | 677 | |||
XM_006233997.2 | XP_006234059.1 | 9, 10 | 676 | |||
XM_006233998.2 | XP_006234060.1 | 9, 10 | 676 | |||
XM_008761774.1 | XP_008759996.1 | 8, 9 | 581 | |||
GenBank mRNA | FN805927.1 | - | 9, 10 | 600 | ||
FQ147221.1 | - | 3, 4 | 141 | |||
U35022.2 | - | 9, 10 | 707 |