Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022713.2 | NP_073204.2 | 4 | 402 | ||
XM_006244202.2 | XP_006244264.1 | 4 | 582 | |||
GenBank mRNA | BC066665.1 | - | 4 | 391 | 5623927 | |
DN934649.1 | - | 4 | 433 | |||
M32725.1 | - | 1 | 43 |