Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003137.4 | NP_003128.3 | 4 | 416 | ||
NR_034069.1 | 4 | 444 | ||||
GenBank mRNA | AJ318054.1 | - | 3 | 814 | ||
AK292277.1 | - | 4 | 361 | |||
AK299591.1 | - | 4 | 404 | |||
AK312712.1 | - | 4 | 308 | |||
BC038292.1 | - | 4 | 381 | 4824261 | ||
BG772464.1 | - | 4 | 424 | |||
BX648663.1 | - | 4 | 333 | |||
BX648697.1 | - | 4 | 334 | |||
DB233331.1 | - | 4 | 416 |