Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hrsp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031714.1 | NP_113902.1 | 5 | 398 | ||
GenBank mRNA | AF015949.1 | - | 5 | 398 | ||
BC078779.1 | - | 5 | 389 | 7128728 | ||
D49363.1 | - | 5 | 368 | |||
FQ209529.1 | - | 5 | 411 | |||
FQ209945.1 | - | 4 | 248 | |||
FQ210021.1 | - | 5 | 403 | |||
FQ218425.1 | - | 5 | 405 | |||
FQ218811.1 | - | 5 | 406 | |||
FQ219440.1 | - | 5 | 405 | |||
FQ219496.1 | - | 5 | 411 | |||
X70825.1 | - | 5 | 366 |