Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031000.3 | NP_112262.1 | 5 | 447 | ||
GenBank mRNA | BC059133.1 | - | 5 | 726 | 6918695 | |
D10854.1 | - | 5 | 447 | |||
FQ212523.1 | - | 5 | 689 | |||
FQ212586.1 | - | 5 | 687 | |||
FQ212919.1 | - | 4 | 397 | |||
FQ213220.1 | - | 4 | 394 | |||
FQ213491.1 | - | 5 | 720 | |||
FQ213579.1 | - | 5 | 689 | |||
FQ213776.1 | - | 5 | 689 | |||
FQ214173.1 | - | 5 | 689 | |||
FQ214441.1 | - | 5 | 466 | |||
FQ214642.1 | - | 704 | ||||
FQ214700.1 | - | 5 | 662 | |||
FQ215620.1 | - | 5 | 514 | |||
FQ215772.1 | - | 5 | 689 | |||
FQ218383.1 | - | 5 | 690 | |||
FQ218721.1 | - | 5 | 689 | |||
FQ220055.1 | - | 3 | 290 | |||
FQ226221.1 | - | 5 | 722 | |||
FQ228132.1 | - | 5 | 722 | |||
FQ228842.1 | - | 5 | 718 | |||
FQ229180.1 | - | 4 | 361 |