Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tas2r121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023997.1 | NP_076487.1 | 1 | 82 | ||
GenBank mRNA | AF227144.1 | - | 1 | 82 |