Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005419.3 | NP_005410.1 | 22 | 2292 | ||
NM_198332.1 | NP_938146.1 | 22 | 2280 | |||
XM_011538697.2 | XP_011536999.1 | 22 | 2224 | |||
XM_011538698.2 | XP_011537000.1 | 22 | 2185 | |||
XM_011538700.2 | XP_011537002.1 | 16 | 1622 | |||
XM_017019904.1 | XP_016875393.1 | 21 | 2659 | |||
GenBank mRNA | AK296939.1 | - | 22 | 2153 | ||
BC051284.1 | - | 21 | 2112 | 6460486 | ||
BX640607.1 | - | 9 | 1019 | |||
M97934.1 | - | 22 | 2126 |