Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243835.1 | NP_001230764.1 | 21, 22 | 2306 | ||
NM_003151.3 | NP_003142.1 | 21, 22 | 2357 | |||
XM_006712719.3 | XP_006712782.1 | 21, 22 | 2352 | |||
XM_011511705.2 | XP_011510007.1 | 24, 25 | 2503 | |||
GenBank mRNA | AK299976.1 | - | 18, 19 | 1838 | ||
AK301172.1 | - | 19, 20 | 1781 | |||
BC031212.1 | - | 21, 22 | 2278 | 4830583 | ||
L78440.1 | - | 21, 22 | 2123 |