Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lum | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031050.1 | NP_112312.1 | 2 | 741 | ||
GenBank mRNA | FQ221670.1 | - | 2 | 808 | ||
FQ229097.1 | - | 2 | 804 | |||
FQ230118.1 | - | 1 | 570 | |||
X84039.1 | - | 2 | 741 |