Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037343.1 | NP_001032420.1 | 13 | 2092 | ||
NM_001323289.1 | NP_001310218.1 | 11 | 2005 | |||
NM_003159.2 | NP_003150.1 | 12 | 2096 | |||
GenBank mRNA | AK307444.1 | - | 12 | 2093 | ||
AY217744.1 | - | 12 | 2064 | |||
HQ171445.1 | - | 11 | 1843 | |||
X89059.1 | - | 1 | 829 | |||
Y15057.1 | - | 12 | 2064 |