Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000455.4 | NP_000446.1 | 6 | 1863 | ||
XM_005259617.2 | XP_005259674.1 | 6 | 1373 | |||
XM_005259618.3 | XP_005259675.1 | 6 | 1373 | |||
XM_011528209.1 | XP_011526511.1 | 7 | 1504 | |||
GenBank mRNA | AB209553.1 | - | 6 | 838 | ||
AF035625.1 | - | 6 | 1086 | |||
AF217978.1 | - | 7 | 1709 | |||
AK314858.1 | - | 6 | 874 | |||
BC007981.2 | - | 6 | 1198 | 3689780 | ||
BC019334.1 | - | 6 | 1019 | 3946005 | ||
EF644172.1 | - | 6 | 832 | |||
KU178339.1 | - | 6 | 748 | |||
U63333.1 | - | 6 | 748 |