Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmsb4x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031136.1 | NP_112398.1 | 3 | 497 | ||
GenBank mRNA | BC058137.1 | - | 3 | 511 | 6889807 | |
BC097308.1 | - | 3 | 496 | |||
FQ211950.1 | - | 3 | 507 | |||
FQ229728.1 | - | 3 | 505 | |||
M34043.1 | - | 3 | 497 |