Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031333.1 | NP_112623.1 | 9 | 1368 | ||
XM_006254441.1 | XP_006254503.1 | 8 | 1734 | |||
GenBank mRNA | AB017695.1 | - | 9 | 1368 | ||
AF097593.1 | - | 8 | 1068 |