Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cish | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031804.1 | NP_113992.1 | 3 | 533 | ||
GenBank mRNA | AF065161.1 | - | 3 | 415 | ||
BC161930.1 | - | 3 | 533 | 8375557 |