Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULT1C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001056.3 | NP_001047.1 | 5 | 933 | ||
NM_176825.2 | NP_789795.1 | 6 | 966 | |||
XM_005264013.3 | XP_005264070.1 | 5 | 754 | |||
GenBank mRNA | AB008164.1 | - | 5 | 603 | ||
AF026303.1 | - | 5 | 565 | |||
AF186251.1 | - | 5 | 798 | |||
AF186252.1 | - | 5 | 933 | |||
AF186253.1 | - | 5 | 649 | |||
AF186254.1 | - | 6 | 831 | |||
AF186255.1 | - | 6 | 966 | |||
AF186256.1 | - | 6 | 682 | |||
AK297086.1 | - | 5 | 739 | |||
AK298306.1 | - | 5 | 604 | |||
AK313193.1 | - | 5 | 697 | |||
AL832145.1 | - | 1 | 5707 | |||
BC005353.1 | - | 5 | 550 | 4051264 | ||
BT006951.1 | - | 4 | 480 | |||
U66036.1 | - | 5 | 656 |