Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031828.1 | NP_114016.1 | 23 | 2908 | ||
GenBank mRNA | AF083341.1 | - | 21 | 2161 | ||
AF135265.1 | - | 21 | 2579 | |||
AY330290.1 | - | 20 | 2038 | |||
AY330291.1 | - | 20 | 2038 | |||
AY330292.1 | - | 20 | 2047 | |||
AY330293.2 | - | 20 | 2038 | |||
AY330294.1 | - | 20 | 2038 | |||
AY344964.1 | - | 18 | 1874 | |||
AY344965.2 | - | 19 | 2450 | |||
U40603.1 | - | 23 | 2908 | |||
U55995.1 | - | 20 | 2191 | |||
U93052.1 | - | 20 | 2139 |