Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt2b15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004271.1 | NP_001004271.1 | 1 | 376 | ||
XM_008770060.1 | XP_008768282.1 | 1 | 403 | |||
GenBank mRNA | BC078782.1 | - | 1 | 376 | 7107952 | |
U06273.1 | - | 1 | 357 | |||
U06274.1 | - | 1 | 358 |