Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANK3 SIRNA Assays ›
Gene Symbol:
ANK3
Gene Name:
ankyrin 3, node of Ranvier (ankyrin G)
Gene Aliases:
ANKYRIN-G, MRT37
Chromosome Location:
Chr.10: 60026298 - 60733526 on Build GRCh38
UniGene ID:
Hs.499725
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s1368
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANK3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001149.3
NP_001140.2
7
1405
NM_001204403.1
NP_001191332.1
30
3543
NM_001204404.1
NP_001191333.1
30
3786
NM_001320874.1
NP_001307803.1
29
3800
NM_020987.4
NP_066267.2
29
3800
XM_005269715.3
XP_005269772.1
30
3687
XM_006717796.3
XP_006717859.1
31
3699
XM_006717802.3
XP_006717865.1
31
3699
XM_011539708.2
XP_011538010.1
31
3699
XM_011539709.2
XP_011538011.1
31
3699
XM_017016110.1
XP_016871599.1
31
3699
XM_017016111.1
XP_016871600.1
30
3687
XM_017016112.1
XP_016871601.1
31
3684
XM_017016113.1
XP_016871602.1
31
3699
XM_017016114.1
XP_016871603.1
31
3798
XM_017016115.1
XP_016871604.1
31
3528
XM_017016116.1
XP_016871605.1
31
3699
XM_017016117.1
XP_016871606.1
31
3699
XM_017016118.1
XP_016871607.1
31
3699
XM_017016119.1
XP_016871608.1
31
3699
XM_017016120.1
XP_016871609.1
31
3699
XM_017016121.1
XP_016871610.1
31
3699
XM_017016122.1
XP_016871611.1
29
3633
XM_017016123.1
XP_016871612.1
31
3699
XM_017016124.1
XP_016871613.1
31
3699
XM_017016125.1
XP_016871614.1
30
3672
XM_017016126.1
XP_016871615.1
29
3567
XM_017016127.1
XP_016871616.1
29
3567
XM_017016128.1
XP_016871617.1
31
3699
XM_017016129.1
XP_016871618.1
31
3699
XM_017016130.1
XP_016871619.1
29
3633
XM_017016131.1
XP_016871620.1
29
3618
XM_017016132.1
XP_016871621.1
29
3633
XM_017016133.1
XP_016871622.1
29
3732
XM_017016134.1
XP_016871623.1
29
3567
XM_017016135.1
XP_016871624.1
29
3732
XM_017016136.1
XP_016871625.1
31
3699
XM_017016137.1
XP_016871626.1
31
3699
XM_017016138.1
XP_016871627.1
31
3699
XM_017016139.1
XP_016871628.1
29
3633
XM_017016140.1
XP_016871629.1
29
3633
XM_017016141.1
XP_016871630.1
29
3567
XM_017016142.1
XP_016871631.1
28
3555
XM_017016143.1
XP_016871632.1
29
3567
XM_017016144.1
XP_016871633.1
29
3567
XM_017016145.1
XP_016871634.1
28
3555
XM_017016146.1
XP_016871635.1
28
3555
XM_017016147.1
XP_016871636.1
28
3555
XM_017016148.1
XP_016871637.1
29
3449
XM_017016149.1
XP_016871638.1
28
3654
XM_017016150.1
XP_016871639.1
29
3633
GenBank mRNA
AB209308.1
-
30
3688
AK092527.1
-
7
1410
AK126851.1
-
11
1488
AK292927.1
-
7
1291
AK295661.1
-
30
3493
AL136710.1
-
7
1064
BX537917.1
-
30
3414
BX648574.1
-
30
3508
U13616.1
-
29
3546
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
mitotic cytokinesis
ER to Golgi vesicle-mediated transport
plasma membrane organization
cytoskeletal anchoring at plasma membrane
signal transduction
axonogenesis
axon guidance
neuromuscular junction development
positive regulation of gene expression
positive regulation of cell communication by electrical coupling
positive regulation of sodium ion transport
magnesium ion homeostasis
neuronal action potential
positive regulation of homotypic cell-cell adhesion
Golgi to plasma membrane protein transport
regulation of potassium ion transport
establishment of protein localization
positive regulation of membrane potential
cellular response to magnesium ion
membrane assembly
protein localization to plasma membrane
maintenance of protein location in plasma membrane
protein targeting to plasma membrane
positive regulation of protein targeting to membrane
positive regulation of membrane depolarization during cardiac muscle cell action potential
negative regulation of delayed rectifier potassium channel activity
positive regulation of sodium ion transmembrane transporter activity
positive regulation of cation channel activity
structural constituent of cytoskeleton
protein binding
cytoskeletal protein binding
cytoskeletal adaptor activity
spectrin binding
protein binding, bridging
ion channel binding
cadherin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0