Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105740.1 | NP_001099210.1 | 1244 | |||
XM_008770476.1 | XP_008768698.1 | 3 | 1408 | |||
XM_008770477.1 | XP_008768699.1 | 3 | 1408 | |||
XM_008770478.1 | XP_008768700.1 | 3 | 1408 | |||
XM_008770479.1 | XP_008768701.1 | 3 | 1408 |
Gene Symbol: NEWGENE_1304700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251622.2 | XP_006251684.1 | 3 | 1408 | ||
XM_006251623.2 | XP_006251685.1 | 3 | 1408 | |||
XM_008770578.1 | XP_008768800.1 | 3 | 1408 | |||
XM_008770579.1 | XP_008768801.1 | 3 | 1408 |