Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cml5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080884.1 | NP_543160.1 | 2 | 955 | ||
XM_006236720.2 | XP_006236782.1 | 2 | 834 | |||
XM_006236721.2 | XP_006236783.1 | 2 | 2363 | |||
XM_006236722.2 | XP_006236784.1 | 2 | 860 | |||
XM_006236723.1 | XP_006236785.1 | 1 | 599 | |||
XM_006236725.2 | XP_006236787.1 | 2 | 618 | |||
XM_008763044.1 | XP_008761266.1 | 3 | 922 | |||
GenBank mRNA | AF187100.1 | - | 1 | 287 | ||
BC158648.1 | - | 2 | 955 | 8367103 |