Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pax3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053710.1 | NP_446162.1 | 7 | 1471 | ||
XM_006245131.2 | XP_006245193.1 | 7 | 1525 | |||
XM_008767181.1 | XP_008765403.1 | 8 | 1604 |