Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TALDO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006755.1 | NP_006746.1 | 3 | 304 | ||
GenBank mRNA | AK295537.1 | - | 3 | 272 | ||
AK313427.1 | - | 3 | 272 | |||
BC010103.2 | - | 3 | 309 | 3536661 | ||
BC018847.2 | - | 3 | 252 | |||
L19437.2 | - | 3 | 304 |