Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003192.2 | NP_003183.1 | 1 | 725 | ||
GenBank mRNA | AK312681.1 | - | 1 | 667 | ||
BC020170.1 | - | 1 | 665 | 3536688 | ||
BE795945.1 | - | 1 | 725 | |||
BT007002.1 | - | 1 | 652 | |||
U61234.1 | - | 1 | 675 |