Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172085.1 | NP_001165556.1 | 5, 6 | 916 | ||
NM_003194.4 | NP_003185.1 | 6, 7 | 1118 | |||
GenBank mRNA | AK298272.1 | - | 7, 8 | 1000 | ||
AK304648.1 | - | 5, 6 | 903 | |||
AK315222.1 | - | 6, 7 | 1081 | |||
BC109053.1 | - | 6, 7 | 1029 | 40032492 | ||
BC109054.1 | - | 7, 8 | 883 | 40032493 | ||
BC110341.1 | - | 6, 7 | 1077 | 5761426 | ||
BT019657.1 | - | 5, 6 | 830 | |||
CR456776.1 | - | 5, 6 | 839 | |||
M34960.1 | - | 5, 6 | 839 | |||
M55654.1 | - | 6, 7 | 1080 | |||
X54993.1 | - | 5, 6 | 925 |