Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053918.2 | NP_446370.2 | 3 | 204 | ||
GenBank mRNA | BC063160.1 | - | 3 | 208 | 6922315 | |
D00575.1 | - | 3 | 178 | |||
J00757.1 | - | 2 | 163 |