Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172317.2 | NP_758528.1 | 3, 4 | 101 | ||
XM_006228818.2 | XP_006228880.1 | 3, 4 | 351 | |||
XM_006228819.2 | XP_006228881.1 | 3, 4 | 205 | |||
XM_006228820.2 | XP_006228882.1 | 3, 4 | 341 | |||
GenBank mRNA | AW142066.1 | - | 2, 3 | 106 | ||
DY471861.1 | - | 3, 4 | 473 |